1,1-Bis(methylthio)-2-nitroethylen
Lieferant: Molekula
|
Achtung
|
M13679748
MOLEM13679748EA
0
EUR
MOLEM13679748
1,1-Bis(methylthio)-2-nitroethylen
1,1-Bis(methylthio)-2-nitroethylen
Formel:
C₄H₇NO₂S₂ Molecular Weight: 165,24 g/mol Schmelzpunkt: 122…125 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00010443 CAS-Nummer: 13623-94-4 EINECS: 237-108-4 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...