Cysteaminiumchlorid

Lieferant: Molekula

M13886169
MOLEM13886169EA 0 EUR
MOLEM13886169
Cysteaminiumchlorid
Cysteaminiumchlorid
Formel: C₂H₇NS·HCl
MDL: MFCD00012904
CAS-Nummer: 156-57-0
EINECS: 205-858-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR