4-Brombenzolsulfonamid

Lieferant: Molekula

Synonyme: 4-Brombenzolsulfonsäureamid

M14162215
MOLEM14162215EA 0 EUR
MOLEM14162215
4-Brombenzolsulfonamid
4-Brombenzolsulfonamid
Formel: C₆H₆BrNO₂S
Molecular Weight: 236,09 g/mol
MDL: MFCD00051977
CAS-Nummer: 701-34-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR