Diosmin

Lieferant: Molekula

Synonyme: 5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one

M14577662
MOLEM14577662EA 0 EUR
MOLEM14577662
Diosmin
Diosmin
Formel: C₂₈H₃₂O₁₅
MDL: MFCD00009772
CAS-Nummer: 520-27-4
EINECS: 208-289-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR