Dodecyltrimethylammoniumbromid
Lieferant: Molekula
Synonyme:
Dodecyl trimethyl ammonium bromide, (1-Dodecyl)trimethylammonium bromide
M14637765
MOLEM14637765EA
0
EUR
MOLEM14637765
Dodecyltrimethylammoniumbromid
Dodecyltrimethylammonium bromide
Formel:
C₁₅H₃₄BrN Schmelzpunkt: 264…266 °C |
MDL:
MFCD00011767 CAS-Nummer: 1119-94-4 EINECS: 214-290-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...