Hesperidin
Lieferant: Molekula
Synonyme:
(S)-5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)chroman-4-one
M14809891
MOLEM14809891EA
0
EUR
MOLEM14809891
Hesperidin
Hesperidin
Formel:
C₂₈H₃₄O₁₅ Molecular Weight: 610,56 g/mol |
MDL:
MFCD00075663 CAS-Nummer: 520-26-3 EINECS: 208-288-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...