1-Nonansulfonsäure Natriumsalz

Lieferant: Molekula

M15204747
MOLEM15204747EA 0 EUR
MOLEM15204747
1-Nonansulfonsäure Natriumsalz
1-Nonanesulphonic acid sodium salt
Formel: C₉H₁₉NaO₃S
MDL: MFCD00025010
CAS-Nummer: 35192-74-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR