4-Phenylazophenol

Lieferant: Molekula

M15523043
MOLEM15523043EA 0 EUR
MOLEM15523043
4-Phenylazophenol
4-Phenylazophenol
Formel: C₁₂H₁₀N₂O
MDL: MFCD00002330
CAS-Nummer: 1689-82-3
EINECS: 216-880-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR