Bathocuproinsulfonat Dinatriumsalz
Lieferant: Molekula
Synonyme:
Bathocuproinedisulphonic Acid Disodium Salt, Disodium Bathocuproinedisulphonate, Bathocuproine disulphonic acid disodium salt
M15611838
MOLEM15611838EA
0
EUR
MOLEM15611838
Bathocuproinsulfonat Dinatriumsalz
Bathocuproinsulfonat Dinatriumsalz
Formel:
C₂₆H₁₈N₂Na₂O₆S₂ |
MDL:
MFCD00011679 CAS-Nummer: 52698-84-7 EINECS: 258-111-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...