4-Hydroxyindol
Lieferant: Molekula
|
Achtung
|
Synonyme:
Indol-4-ol, 4-Hydroxy-1H-indol, 4-Indolol, 1H-Indol-4-ol
M15874512
MOLEM15874512EA
0
EUR
MOLEM15874512
4-Hydroxyindol
4-Hydroxyindol
Formel:
C₈H₇NO Molecular Weight: 133,15 g/mol Siedepunkt: 100 °C (0,01 mmHg) Schmelzpunkt: 102…104 °C |
MDL:
MFCD00005667 CAS-Nummer: 2380-94-1 EINECS: 219-177-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...