(R)-(+)-Hydroxypyrrolidin

Lieferant: Molekula

Synonyme: (R)-Pyrrolidin-3-ol, (R)-(+)-3-Pyrrolidinol, (R)-(+)-3-Hydroxypyrrolidine

M15970045
MOLEM15970045EA 0 EUR
MOLEM15970045
(R)-(+)-Hydroxypyrrolidin
(R)-(+)-Hydroxypyrrolidin
Formel: C₄H₉NO
Molecular Weight: 87.12 g/mol
MDL: MFCD00145220
CAS-Nummer: 2799-21-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR