Methyl-5-methylisoxazol-3-carboxylat
Lieferant: Molekula
Synonyme:
Methyl 5-methylisoxazole-3-carboxylate
M16123593
MOLEM16123593EA
0
EUR
MOLEM16123593
Methyl-5-methylisoxazol-3-carboxylat
Methyl-5-methylisoxazol-3-carboxylat
Formel:
C₆H₇NO₃ Molecular Weight: 141,13 g/mol |
MDL:
MFCD00015895 CAS-Nummer: 19788-35-3 EINECS: 243-311-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...