N-Acetyl-DL-methionin

Lieferant: Molekula

Synonyme: Ac-DL-Met-OH

M16190196
MOLEM16190196EA 0 EUR
MOLEM16190196
N-Acetyl-DL-methionin
N-Acetyl-DL-methionin
Formel: C₇H₁₃NO₃S
Molecular Weight: 191,2 g/mol
MDL: MFCD00008681
CAS-Nummer: 1115-47-5
EINECS: 214-224-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR