2,8-Chinolindiol

Lieferant: Molekula
Achtung

Synonyme: 2,8-Dihydroxyquinoline

M16200530
MOLEM16200530EA 0 EUR
MOLEM16200530
2,8-Chinolindiol
2,8-Chinolindiol
Formel: C₉H₇NO₂
Molecular Weight: 161.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00216696
CAS-Nummer: 15450-76-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR