4-Brom-3,5-dihydroxybenzoesäure

Lieferant: Molekula
Achtung

M16601852
MOLEM16601852EA 0 EUR
MOLEM16601852
4-Brom-3,5-dihydroxybenzoesäure
4-Bromo-3,5-dihydroxybenzoic acid
Formel: C₇H₅BrO₄
Molecular Weight: 233,02 g/mol
MDL: MFCD00002513
CAS-Nummer: 16534-12-6
EINECS: 240-606-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR