Kaliumselenocyanat

Lieferant: Molekula
Gefahr

M16609554
MOLEM16609554EA 0 EUR
MOLEM16609554
Kaliumselenocyanat
Potassium selenocyanate
Formel: CKNSe
Molecular Weight: 144,08 g/mol
MDL: MFCD00011366
CAS-Nummer: 3425-46-5
EINECS: 222-320-1
UN: 3283
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR