Daidzein

Lieferant: Molekula

Synonyme: 7-Hydroxy-3-(4-hydroxyphenyl)chromen-4-on, 7-Hydroxy-3-(4-hydroxyphenyl)-4-benzopyron, 4',7-Dihydroxyisoflavon, Daidzeol

M17127849
MOLEM17127849EA 0 EUR
MOLEM17127849
Daidzein
Daidzein
Formel: C₁₅H₁₀O₄
Molecular Weight: 254.24 g/mol
Siedepunkt: 513
Schmelzpunkt: 212…214 °C
MDL: MFCD00016954
CAS-Nummer: 486-66-8
EINECS: 207-635-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR