Curcumin

Lieferant: Molekula

Synonyme: (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione

M17132282
MOLEM17132282EA 0 EUR
MOLEM17132282
Curcumin
Curcumin
Formel: C₂₁H₂₀O₆
Molecular Weight: 368,39 g/mol
MDL: MFCD00008365
CAS-Nummer: 458-37-7
EINECS: 207-280-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR