N,O-Dimethylhydroxylamin Hydrochlorid

Lieferant: Molekula

M17470360
MOLEM17470360EA 0 EUR
MOLEM17470360
N,O-Dimethylhydroxylamin Hydrochlorid
N,O-Dimethylhydroxylamin Hydrochlorid
Formel: C₂H₈ClNO
MDL: MFCD00012485
CAS-Nummer: 6638-79-5
EINECS: 229-642-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR