3-Methylcrotonsäure

Lieferant: Molekula

Synonyme: 3,3-Dimethylacrylic acid, 3-Methylbut-2-enoic acid

M17476017
MOLEM17476017EA 0 EUR
MOLEM17476017
3-Methylcrotonsäure
3-Methylcrotonsäure
Formel: C₅H₈O₂
Molecular Weight: 100,12 g/mol
MDL: MFCD00004366
CAS-Nummer: 541-47-9
EINECS: 208-782-7
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR