2,4-Dimethylthiazol

Lieferant: Molekula

M17533338
MOLEM17533338EA 0 EUR
MOLEM17533338
2,4-Dimethylthiazol
2,4-Dimethylthiazole
Formel: C₅H₇NS
Molecular Weight: 113,18 g/mol
Siedepunkt: 144 °C (1013 hPa)
Dichte: 1,0601 g/cm³ (15 °C)
MDL: MFCD00014509
CAS-Nummer: 541-58-2
EINECS: 208-786-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR