2,4-Dichlorpyrimidin
Lieferant: Molekula
|
Achtung
|
M17839790
MOLEM17839790EA
0
EUR
MOLEM17839790
2,4-Dichlorpyrimidin
2,4-Dichloropyrimidine
Formel:
C₄H₂Cl₂N₂ Molecular Weight: 148,98 g/mol Siedepunkt: 209 °C (1013 hPa) Schmelzpunkt: 61 °C |
MDL:
MFCD00006061 CAS-Nummer: 3934-20-1 EINECS: 223-508-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...