3-Indolcarbonsäure

Lieferant: Molekula

Synonyme: 1H-Indole-3-carboxylic acid

M17920095
MOLEM17920095EA 0 EUR
MOLEM17920095
3-Indolcarbonsäure
3-Indolcarbonsäure
Formel: C₉H₇NO₂
MDL: MFCD00005624
CAS-Nummer: 771-50-6
EINECS: 212-231-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR