5-Bromuracil
Lieferant: Molekula
Synonyme:
Bromuracil, 5-Brom-2,4-dihydroxypyrimidin
M18193444
MOLEM18193444EA
0
EUR
MOLEM18193444
5-Bromuracil
5-Bromuracil
Formel:
C₄H₃BrN₂O₂ Molecular Weight: 190,98 g/mol Siedepunkt: 384 °C (1013 hPa) Schmelzpunkt: 165 °C |
MDL:
MFCD00006017 CAS-Nummer: 51-20-7 EINECS: 200-084-0 Merck Index: 13,01425 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...