2-Chlor-3-fluoranilin

Lieferant: Molekula

M18483005
MOLEM18483005EA 0 EUR
MOLEM18483005
2-Chlor-3-fluoranilin
2-Chlor-3-fluoranilin
Formel: C₆H₅ClFN
Molecular Weight: 145,56 g/mol
MDL: MFCD03407961
CAS-Nummer: 21397-08-0
EINECS: 244-366-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR