1,2,3-Triazol
Lieferant: Molekula
Synonyme:
1,2,3-1H-Triazol
M18484185
MOLEM18484185EA
0
EUR
MOLEM18484185
1,2,3-Triazol
1,2,3-Triazole
Formel:
C₂H₃N₃ Molecular Weight: 69,07 g/mol Siedepunkt: 208 °C (1013 hPa) Schmelzpunkt: 23…25 °C Dichte: 1,192 g/cm³ (20 °C) |
MDL:
MFCD00014490 CAS-Nummer: 288-36-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...