2-Fluorphenylboronsäure

Lieferant: Molekula

Synonyme: (2-Fluorphenyl)metaborsäure, 2-Fluorbenzolboronsäure, Dihydroxy(2-fluorphenyl)boran

M18550854
MOLEM18550854EA 1290 EUR
MOLEM18550854
2-Fluorphenylboronsäure
2-Fluorphenylboronsäure
Formel: C₆H₆BFO₂
Molecular Weight: 139,92 g/mol
MDL: MFCD00674013
CAS-Nummer: 1993-03-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR