1,2-Bis(diphenylphosphino)ethan

Lieferant: Molekula
Achtung

Synonyme: Ethan-1,3-diylbis(diphenylphosphin), Bis(1,2-diphenylphosphino)ethan, Ethylenbis(diphenylphosphin), DIPHOS

M18770139
MOLEM18770139EA 0 EUR
MOLEM18770139
1,2-Bis(diphenylphosphino)ethan
1,2-Bis(diphenylphosphino)ethan
Formel: C₂₆H₂₄P₂
Molecular Weight: 398,42 g/mol
Schmelzpunkt: 140…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003047
CAS-Nummer: 1663-45-2
EINECS: 216-769-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR