Tryptamin

Lieferant: Molekula

Synonyme: 2-(1H-indol-3-yl)ethan-1-amine

M18963277
MOLEM18963277EA 0 EUR
MOLEM18963277
Tryptamin
Tryptamin
Formel: C₁₀H₁₂N₂
Molecular Weight: 160,22 g/mol
MDL: MFCD00005661
CAS-Nummer: 61-54-1
EINECS: 200-510-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR