5-Amino-4,6-dichlorpyrimidin
Lieferant: Molekula
Synonyme:
4,6-Dichloropyrimidin-5-ylamine
M19100531
MOLEM19100531EA
0
EUR
MOLEM19100531
5-Amino-4,6-dichlorpyrimidin
5-Amino-4,6-dichloropyrimidine
Formel:
C₄H₃Cl₂N₃ Molecular Weight: 163,99 g/mol Siedepunkt: 262 °C (1013 hPa) Schmelzpunkt: 147 °C |
MDL:
MFCD00006108 CAS-Nummer: 5413-85-4 EINECS: 226-503-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...