5-Iod-2'-deoxyuridin (Idoxuridin)

Lieferant: Molekula

M19147388
MOLEM19147388EA 0 EUR
MOLEM19147388
5-Iod-2'-deoxyuridin (Idoxuridin)
5-Iod-2'-deoxyuridin (Idoxuridin)
Formel: C₉H₁₁IN₂O₅
Lagertemperatur: Raumtemperatur
MDL: MFCD00134656
CAS-Nummer: 54-42-2
EINECS: 200-207-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR