Luminol (3-Aminophthalhydrazid)

Lieferant: Molekula

Synonyme: NSC 5064

M19611311
MOLEM19611311EA 181 EUR
MOLEM19611311
Luminol (3-Aminophthalhydrazid)
Luminol (3-Aminophthalhydrazid)
Formel: C₈H₇N₃O₂
Molecular Weight: 177.16 g/mol
MDL: MFCD00006890
CAS-Nummer: 521-31-3
EINECS: 208-309-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR