L(+)-α-Phenylglycin

Lieferant: Molekula

Synonyme: H-Phg-OH, L-(+)-2-Phenylglycine

M20595516
MOLEM20595516EA 0 EUR
MOLEM20595516
L(+)-α-Phenylglycin
L(+)-α-Phenylglycin
Formel: C₈H₉NO₂
Molecular Weight: 151,16 g/mol
MDL: MFCD00064403
CAS-Nummer: 2935-35-5
EINECS: 220-909-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR