D-Alanin

Lieferant: Molekula

Synonyme: (R)-(-)-2-Aminopropionic acid, Alanin, D-α-Alanin, D(-)-Alanin, (R)-(-)-Alanin, H-D-Ala-OH, α-Alanin, (R)-(-)-2-Aminopropansäure, D(-)-2-Aminopropionic acid, (R)-(-)-2-Aminopropionsäure

M20667745
MOLEM20667745EA 0 EUR
MOLEM20667745
D-Alanin
D-Alanin
Formel: C₃H₇NO₂
Molecular Weight: 89,09 g/mol
Siedepunkt: 189 °C (1013 hPa)
Schmelzpunkt: 79 °C
MDL: MFCD00008077
CAS-Nummer: 338-69-2
EINECS: 206-418-1
Merck Index: 13,00203

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR