D(-)-Leucinol

Lieferant: Molekula

M20692720
MOLEM20692720EA 0 EUR
MOLEM20692720
D(-)-Leucinol
D(-)-Leucinol
Formel: C₆H₁₅NO
Siedepunkt: 199 °C (1013 hPa)
Dichte: 0,897 g/cm³ (20 °C)
MDL: MFCD00004734
CAS-Nummer: 53448-09-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR