5-Nitroindol

Lieferant: Molekula

Synonyme: 5-Nitro-1H-indole

M20819045
MOLEM20819045EA 0 EUR
MOLEM20819045
5-Nitroindol
5-Nitroindol
Formel: C₈H₆N₂O₂
Molecular Weight: 162,15 g/mol
Schmelzpunkt: 140 °C
MDL: MFCD00005673
CAS-Nummer: 6146-52-7
EINECS: 228-153-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR