9-Aminoacridin Hydrochlorid Monohydrat

Lieferant: Molekula

M20982371
MOLEM20982371EA 0 EUR
MOLEM20982371
9-Aminoacridin Hydrochlorid Monohydrat
9-Aminoacridin Hydrochlorid Monohydrat
Formel: C₁₃H₁₁ClN₂·1H₂O
MDL: MFCD00150071
CAS-Nummer: 52417-22-8
EINECS: 205-145-5
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR