L(-)-Mannose

Lieferant: Molekula

Synonyme: (2R,3R,4S,5S)-2,3,4,5,6-Pentahydroxyhexanal, L-(-)-Mannose

M21123215
MOLEM21123215EA 0 EUR
MOLEM21123215
L(-)-Mannose
L(-)-Mannose
Formel: C₆H₁₂O₆
Molecular Weight: 180,16 g/mol
Siedepunkt: 376 °C (1013 hPa)
Schmelzpunkt: 132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00136021
CAS-Nummer: 10030-80-5
EINECS: 233-080-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR