alle-trans-Retinol

Lieferant: Molekula

Synonyme: all-trans-Retinol, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-ol, Vitamin A alcohol

M21418841
MOLEM21418841EA 0 EUR
MOLEM21418841
alle-trans-Retinol
alle-trans-Retinol
Formel: C₂₀H₃₀O
Molecular Weight: 286,46 g/mol
MDL: MFCD00001552
CAS-Nummer: 68-26-8
EINECS: 200-683-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR