5-Chlor-2-mercaptobenzothiazol
Lieferant: Molekula
Synonyme:
5-Chlorobenzo[d]thiazole-2(3H)-thione
M21566757
MOLEM21566757EA
0
EUR
MOLEM21566757
5-Chlor-2-mercaptobenzothiazol
5-Chloro-2-mercaptobenzothiazole
Formel:
C₇H₄ClNS₂ Molecular Weight: 201,7 g/mol |
MDL:
MFCD00005783 CAS-Nummer: 5331-91-9 EINECS: 226-235-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...