4-Azaindol

Lieferant: Molekula

Synonyme: 1H-Pyrrolo[2,3-e]pyridin

M21910645
MOLEM21910645EA 0 EUR
MOLEM21910645
4-Azaindol
4-Azaindol
Formel: C₇H₆N₂
MDL: MFCD00971977
CAS-Nummer: 272-49-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR