Dimethyl-2-aminoterephthalat
Lieferant: Molekula
Synonyme:
Dimethyl aminoterephthalate, 2-Aminodimethyl terephthalate
M22351781
MOLEM22351781EA
0
EUR
MOLEM22351781
Dimethyl-2-aminoterephthalat
Dimethyl-2-aminoterephthalat
Formel:
C₁₀H₁₁NO₄ Molecular Weight: 209.2 g/mol |
MDL:
MFCD00008427 CAS-Nummer: 5372-81-6 EINECS: 226-364-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...