3-Chlorsalicylsäure
Lieferant: Molekula
M22382330
MOLEM22382330EA
228
EUR
MOLEM22382330
3-Chlorsalicylsäure
3-Chlorsalicylsäure
Formel:
C₇H₅ClO₃ Molecular Weight: 172,57 g/mol Siedepunkt: 320 °C (1013 hPa) Schmelzpunkt: 183…183,5 °C |
MDL:
MFCD00010390 CAS-Nummer: 1829-32-9 EINECS: 217-379-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...