1,2,3,4-Tetrahydronaphthalin-1-on (1-Tetralon, α-Tetralon)

Lieferant: Molekula

Synonyme: 3,4-Dihydronaphthalen-1(2H)-one

M22571232
MOLEM22571232EA 0 EUR
MOLEM22571232
1,2,3,4-Tetrahydronaphthalin-1-on (1-Tetralon, α-Tetralon)
1,2,3,4-Tetrahydronaphthalin-1-on (1-Tetralon, α-Tetralon)
Formel: C₁₀H₁₀O
Molecular Weight: 146,19 g/mol
MDL: MFCD00001688
CAS-Nummer: 529-34-0
EINECS: 208-460-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR