Benzo-15-krone-5 (Benzo-1,4,7,10,13-pentaoxacyclopentadecan)

Lieferant: Molekula

Synonyme: Benzo-15-crown-5

M22590378
MOLEM22590378EA 0 EUR
MOLEM22590378
Benzo-15-krone-5 (Benzo-1,4,7,10,13-pentaoxacyclopentadecan)
Benzo-15-krone-5 (Benzo-1,4,7,10,13-pentaoxacyclopentadecan)
Formel: C₁₄H₂₀O₅
Molecular Weight: 268.31 g/mol
MDL: MFCD00005945
CAS-Nummer: 14098-44-3
EINECS: 237-947-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR