L(+)-Cystein Hydrochlorid

Lieferant: Molekula

Synonyme: H-Cys-OH.HCl, (R)-(+)-Cystein Hydrochlorid, (R)-(+)-2-Amino-3-mercaptopropansäure Hydrochlorid

M22842789
MOLEM22842789EA 0 EUR
MOLEM22842789
L(+)-Cystein Hydrochlorid
L(+)-Cystein Hydrochlorid
Formel: HSCH₂CH(NH₂)COOH·HCl
Molecular Weight: 157.62 g/mol
Schmelzpunkt: 175
Lagertemperatur: Raumtemperatur
MDL: MFCD00064553
CAS-Nummer: 52-89-1
EINECS: 200-157-7
Merck Index: 13,02810

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR