D(-)-allo-Isoleucin
Lieferant: Molekula
Synonyme:
(2R,3S)-(-)-α-Amino-β-methylvaleriansäure, (2R,3S)-(-)-allo-Isoleucin, (2R,3S)-(-)-2-Amino-3-methylpentansäure, H-D-allo-Ile-OH
M23313421
MOLEM23313421EA
0
EUR
MOLEM23313421
D(-)-allo-Isoleucin
D(-)-allo-Isoleucin
Formel:
C₆H₁₃NO₂ Molecular Weight: 131,17 g/mol |
MDL:
MFCD00066445 CAS-Nummer: 1509-35-9 EINECS: 216-143-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...