4-(Phenylazo)diphenylamin

Lieferant: Molekula

Synonyme: 4-Phenylazodiphenylamine

M23320388
MOLEM23320388EA 0 EUR
MOLEM23320388
4-(Phenylazo)diphenylamin
4-(Phenylazo)diphenylamin
Formel: C₁₈H₁₅N₃
Molecular Weight: 273,34 g/mol
MDL: MFCD00003023
CAS-Nummer: 101-75-7
EINECS: 202-972-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR