1-Aza-15-krone-5 (1,4,7,10-Tetraoxa-13-azacyclopentadecan)

Lieferant: Molekula

Synonyme: 1,4,7,10-Tetraoxa-13-azacyclopentadecane, 1-Aza-15-crown 5-Ether, 1,4,7,10-Tetraoxa-13-azacyclopentadecane

M23329413
MOLEM23329413EA 0 EUR
MOLEM23329413
1-Aza-15-krone-5 (1,4,7,10-Tetraoxa-13-azacyclopentadecan)
1-Aza-15-krone-5 (1,4,7,10-Tetraoxa-13-azacyclopentadecan)
Formel: C₁₀H₂₁NO₄
Molecular Weight: 219.28 g/mol
MDL: MFCD00075465
CAS-Nummer: 66943-05-3
EINECS: 266-523-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR