4-Brom-2-thiophencarbaldehyd
Lieferant: Molekula
Synonyme:
4-Bromo-2-thiophenecarboxaldehyde, 4-Bromothiophene-2-carboxaldehyde
M23521866
MOLEM23521866EA
0
EUR
MOLEM23521866
4-Brom-2-thiophencarbaldehyd
4-Brom-2-thiophencarbaldehyd
Formel:
C₅H₃BrOS Molecular Weight: 191.05 g/mol |
MDL:
MFCD00005431 CAS-Nummer: 18791-75-8 EINECS: 242-577-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...